(2R,4S)-N-Boc-4-hydroxypiperidine-2-carboxylic acidmethyl ester 95%, 98% ee
Lieferant: Thermo Scientific
Synonyme:
1-tert-Butyl 2-methyl (2R,4S)-4-hydroxypiperidine-1,2-dicarboxylate
362970010
ACRO362970010EA
435
EUR
ACRO362970010
(2R,4S)-N-Boc-4-hydroxypiperidine-2-carboxylic acidmethyl ester 95%, 98% ee
(2R,4S)-N-Boc-4-hydroxypiperidine-2-carboxylic acidmethyl ester
Appearance: Colorless to amber Viscous liquid or waxy mass
Formel:
C₁₂H₂₁NO₅ Molecular Weight: 259,302 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD02259744 CAS-Nummer: 321744-26-7 |
Spezifikation Testergebnisse
Appearance | Colorless to amber Viscous liquid or waxy mass |
Infrared spectrum | Conforms |
HPLC | ≥94 % |
Enantiomeric excess | ≥97.5 % (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...