cis-Dichlorobis(2,2'-bipyridine)ruthenium(II) dihydrate 99%

Lieferant: Thermo Fisher Scientific
Achtung

363300010
ACRO363300010EA 164 EUR
ACRO363300010
cis-Dichlorobis(2,2'-bipyridine)ruthenium(II) dihydrate 99%
cis-Dichlorobis(2,2'-bipyridine)ruthenium(II) dihydrate

Appearance: Black Powder or crystals

Formel: C₂₀H₂₀Cl₂N₄O₂Ru
Lagertemperatur: Raumtemperatur
MDL: MFCD00015504
CAS-Nummer: 15746-57-3

Bestellen

Spezifikation Testergebnisse

Appearance Black Powder or crystals
Infrared spectrum Conforms
Elemental Analysis ≥98.5 % (ex Ru)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR