Acetylacetonatobis(ethylene)rhodium(I) 99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
363381000
ACRO363381000EA
125
EUR
ACRO363381000
Acetylacetonatobis(ethylene)rhodium(I) 99%
Acetylacetonatobis(ethylene)rhodium(I)
Appearance: Orange Crystalline powder or crystals
Formel:
C₉H₁₆O₂Rh Lagertemperatur: Raumtemperatur |
MDL:
MFCD00015354 CAS-Nummer: 12082-47-2 |
Spezifikation Testergebnisse
Appearance | Orange Crystalline powder or crystals |
Infrared spectrum | Conforms |
Elemental Analysis | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...