Bis(tricyclohexylphosphine)palladium(0) 98%
Lieferant: Thermo Scientific
363511000
ACRO363511000EA
81.3
EUR
ACRO363511000
Bis(tricyclohexylphosphine)palladium(0) 98%
Bis(tricyclohexylphosphine)palladium(0)
Appearance: Off-white to light green Powder
Formel:
C₃₆H₆₆P₂Pd Lagertemperatur: Raumtemperatur |
MDL:
MFCD01073796 CAS-Nummer: 33309-88-5 |
Spezifikation Testergebnisse
Appearance | Off-white to light green Powder |
Infrared spectrum | Conforms |
NMR (31P-NMR) | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...