Tetrakis(acetonitrile)palladium(II) tetrafluoroborate 99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
363520010
ACRO363520010EA
184
EUR
ACRO363520010
Tetrakis(acetonitrile)palladium(II) tetrafluoroborate 99%
Appearance: Yellow to green Powder or crystals
Formel:
C₈H₁₂B₂F₈N₄Pd Schmelzpunkt: 230 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00043297 CAS-Nummer: 21797-13-7 |
Spezifikation Testergebnisse
Appearance | Yellow to green Powder or crystals |
Infrared spectrum | Conforms |
Elemental Analysis | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...