(1,5-Cyclooctadiene)(pyridine)(tricyclohexylphosphine)iridium(I) hexafluorophosphate 99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
363581000
363585000
ACRO363581000EA
165
EUR
ACRO363581000
ACRO363585000
(1,5-Cyclooctadiene)(pyridine)(tricyclohexylphosphine)iridium(I) hexafluorophosphate 99%
(1,5-Cyclooctadiene)(pyridine)(tricyclohexylphosphine)iridium(I) hexafluorophosphate
CAS No.: 64536-78-3
Formel:
C₃₁H₅₀F₆IrNP₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00075097 CAS-Nummer: 64536-78-3 |
Spezifikation Testergebnisse
Appearance | Orange Powder |
Infrared spectrum | Conforms |
Carbon (C) | 45.6 to 47.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...