(1,5-Cyclooctadiene)(pyridine)(tricyclohexylphosphine)iridium(I) hexafluorophosphate 99%

Lieferant: Thermo Fisher Scientific
Achtung

363581000 363585000
ACRO363581000EA 165 EUR
ACRO363581000 ACRO363585000
(1,5-Cyclooctadiene)(pyridine)(tricyclohexylphosphine)iridium(I) hexafluorophosphate 99%
(1,5-Cyclooctadiene)(pyridine)(tricyclohexylphosphine)iridium(I) hexafluorophosphate

CAS No.: 64536-78-3

Formel: C₃₁H₅₀F₆IrNP₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00075097
CAS-Nummer: 64536-78-3

Bestellen

Spezifikation Testergebnisse

Appearance Orange Powder
Infrared spectrum Conforms
Carbon (C) 45.6 to 47.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR