2,2'-Bipyridin-4,4'-dicarbonsäure 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 4,4'-Dicarboxy-2,2'-bipyridine, [2,2'-Bipyridine]-4,4'-dicarboxylic acid

363660010
ACRO363660010EA 70.3 EUR
ACRO363660010
2,2'-Bipyridin-4,4'-dicarbonsäure 98%
2,2'-Bipyridin-4,4'-dicarbonsäure
Formel: C₁₂H₈N₂O₄
Molecular Weight: 244.20 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00015430
CAS-Nummer: 6813-38-3

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder or crystals
Infrared spectrum Conforms
Titration excess NaOH ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR