Chlor(1,5-cyclooctadien)iridium(I) dimer
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
[Ir(cod)Cl]2, Chloro-1,5-cyclooctadiene iridium(I) dimer
363962500
363960010
363960050
ACRO363962500EA
115
EUR
ACRO363962500
ACRO363960010
ACRO363960050
Chlor(1,5-cyclooctadien)iridium(I) dimer
Chlor(1,5-cyclooctadien)iridium(I) dimer
Formel:
C₁₆H₂₄Cl₂Ir₂ Molecular Weight: 671.70 g/mol Schmelzpunkt: 190 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012414 CAS-Nummer: 12112-67-3 EINECS: 235-170-7 |
Spezifikation Testergebnisse
Appearance | Orange to red Powder |
Infrared spectrum | Conforms |
Iridium (Ir) | ≥56.4 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...