(2,3-Dichlorphenyl)essigsäure 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2,3-Dichlorophenylacetic acid
365550250
ACRO365550250EA
130
EUR
ACRO365550250
(2,3-Dichlorphenyl)essigsäure 97%
(2,3-Dichlorphenyl)essigsäure
Formel:
(C₆H₃Cl₂)CH₂COOH Molecular Weight: 205,04 g/mol Schmelzpunkt: 130…135 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01861393 CAS-Nummer: 10236-60-9 |
Spezifikation Testergebnisse
Appearance | White Powder |
Infrared spectrum | Conforms |
Melting point | 130 to 135 °C |
HPLC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...