Methyl-3-(trifluoromethyl)phenylacetate 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Methyl 2-(3-(trifluoromethyl)phenyl)acetate
365600050
ACRO365600050EA
58.8
EUR
ACRO365600050
Methyl-3-(trifluoromethyl)phenylacetate 97%
Methyl-3-(trifluoromethyl)phenylacetate
Appearance: Clear colorless Liquid
Formel:
C₁₀H₉F₃O₂ Molecular Weight: 218,175 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04039762 CAS-Nummer: 62451-84-7 |
Spezifikation Testergebnisse
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4500 to 1.4520 (20°C, 589 nm) |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...