DL-Lysin Monohydrat 97%
Lieferant: Thermo Fisher Scientific
365650250
ACRO365650250EA
104
EUR
ACRO365650250
DL-Lysin Monohydrat 97%
DL-Lysin Monohydrat
Formel:
C₆H₁₄N₂O₂·1H₂O |
MDL:
MFCD00151036 CAS-Nummer: 885701-25-7 EINECS: 200-740-6 |
Spezifikation Testergebnisse
Appearance | White to yellow Crystalline powder |
Infrared spectrum | Conforms |
Loss on drying | 7 % to 12 % (105°C, 2 hrs) |
Titration with HClO4 | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...