tert-Butyl-N-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbamate 97+%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: N-Boc-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, 4-(Boc-amino)phenylboronic acid Pinacol ester, tert-Butyl [4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbamate, 4-[(tert-Butoxycarbonyl)amino]phenylboronic acid Pinacol ester, 4-Boc-aminophenylboronic acid, pinacol ester, N-(tert-Butoxycarbonyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, tert-Butyl (4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)carbamate, [4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbamic acid tert-butyl ester, 4-(Boc-amino)benzeneboronic acid pinacol ester

366350010 366350050
ACRO366350010EA 32 EUR
ACRO366350010 ACRO366350050
tert-Butyl-N-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbamate 97+%
tert-Butyl-N-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbamate

Appearance: White to off-white Crystalline powder

Formel: C₁₇H₂₆BNO₄
Schmelzpunkt: 166…169 °C
Lagertemperatur: Kühlschrank
MDL: MFCD02179439
CAS-Nummer: 330793-01-6

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 168 to 173 °C
HPLC ≥97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR