4-Aminophenylboronsäurepinakolester 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
366380010
366380050
ACRO366380010EA
76.6
EUR
ACRO366380010
ACRO366380050
4-Aminophenylboronsäurepinakolester 97%
4-Aminophenylboronsäurepinakolester
Formel:
C₁₂H₁₈BNO₂ Schmelzpunkt: 165…170 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD02093721 CAS-Nummer: 214360-73-3 |
Spezifikation Testergebnisse
Appearance | White to light beige Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 165 to 170 °C |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...