4-Hydroxyphenylboronsäurepinakolester 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
366420050
ACRO366420050EA
93
EUR
ACRO366420050
4-Hydroxyphenylboronsäurepinakolester 97%
4-Hydroxyphenylboronsäurepinakolester
Formel:
C₁₂H₁₇BO₃ Schmelzpunkt: 112…117 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD02093756 CAS-Nummer: 269409-70-3 |
Spezifikation Testergebnisse
Appearance | Yellow to light brown Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 112 to 117 °C |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...