N,N,N'-Trimethylethylendiamin 97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
1-(Dimethylamino)-2-(methylamino)ethan, Dimethyl(2-(methylamino)ethyl)amin
366910050
366910250
366911000
ACRO366910050EA
79.8
EUR
ACRO366910050
ACRO366910250
ACRO366911000
N,N,N'-Trimethylethylendiamin 97%
N,N,N'-Trimethylethylendiamin
Formel:
C₅H₁₄N₂ Molecular Weight: 102.18 g/mol Siedepunkt: 116…118 °C (760 mmHg) Dichte: 0.8 g/cm³ Flammpunkt: 9 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00014874 CAS-Nummer: 142-25-6 EINECS: 205-529-2 UN: 2733 ADR: 3 II |
Spezifikation Testergebnisse
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4180 to 1.4210 (20°C, 589 nm) |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...