N,N,N'-Trimethylethylendiamin 97%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 1-(Dimethylamino)-2-(methylamino)ethan, Dimethyl(2-(methylamino)ethyl)amin

366910050 366910250 366911000
ACRO366910050EA 79.8 EUR
ACRO366910050 ACRO366910250 ACRO366911000
N,N,N'-Trimethylethylendiamin 97%
N,N,N'-Trimethylethylendiamin
Formel: C₅H₁₄N₂
Molecular Weight: 102.18 g/mol
Siedepunkt: 116…118 °C (760 mmHg)
Dichte: 0.8 g/cm³
Flammpunkt: 9 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00014874
CAS-Nummer: 142-25-6
EINECS: 205-529-2
UN: 2733
ADR: 3 II

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4180 to 1.4210 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR