1-(2-Methoxyethyl)piperazin 98%

Lieferant: Thermo Fisher Scientific
Gefahr

367110010
ACRO367110010EA 37.4 EUR
ACRO367110010
1-(2-Methoxyethyl)piperazin 98%
1-(2-Methoxyethyl)piperazin
Formel: C₇H₁₆N₂O
Flammpunkt: 87 °C
MDL: MFCD00191214
CAS-Nummer: 13484-40-7
UN: 2735
ADR: 8,III

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4720 to 1.4750 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR