5'-Brom-2'-hydroxyacetophenon 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
1-(5-Brom-2-hydroxyphenyl)ethan-1-on, 2-Acetyl-4-bromphenol
367210100
ACRO367210100EA
38.2
EUR
ACRO367210100
5'-Brom-2'-hydroxyacetophenon 98%
5'-Brom-2'-hydroxyacetophenon
Formel:
C₈H₇BrO₂ Schmelzpunkt: 58…61 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00191850 CAS-Nummer: 1450-75-5 |
Spezifikation Testergebnisse
Appearance | White to yellow to beige Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 57 to 61 °C |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...