2-Aminoethyldihydrogenphosphat 98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: O-Phosphorylethanolamine

367420050 367420250 367421000
ACRO367420050EA 38.1 EUR
ACRO367420050 ACRO367420250 ACRO367421000
2-Aminoethyldihydrogenphosphat 98%
2-Aminoethyldihydrogenphosphat
Formel: C₂H₈NO₄P
Molecular Weight: 141.0630 g/mol
Schmelzpunkt: 241…243 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00008178
CAS-Nummer: 1071-23-4
EINECS: 213-988-5
UN: 3261
ADR: 8 II

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Powder
Infrared spectrum Conforms
Titration with HClO4 ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR