4-(Ethoxycarbonyl)phenylboronsäure 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 4-(Ethoxycarbonyl)benzeneboronic acid, (4-Ethoxycarbonylphenyl)boronic acid

367790010 367790050 367790250
ACRO367790010EA 54.4 EUR
ACRO367790010 ACRO367790050 ACRO367790250
4-(Ethoxycarbonyl)phenylboronsäure 97%
4-(Ethoxycarbonyl)phenylboronsäure
Formel: C₉H₁₁BO₄
Schmelzpunkt: 184…189 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02179441
CAS-Nummer: 4334-88-7

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder
Infrared spectrum Conforms
Titration with NaOH ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR