N-(tert-Butoxycarbonyl)-L-tert-leucine 98%

Lieferant: Thermo Fisher Scientific

368010010 368010050 368010250
ACRO368010010EA 33.5 EUR
ACRO368010010 ACRO368010050 ACRO368010250
N-(tert-Butoxycarbonyl)-L-tert-leucine 98%
N-(tert-Butoxycarbonyl)-L-tert-leucine

CAS No.: 62965-35-9

Formel: C₁₁H₂₁NO₄
Schmelzpunkt: 118…121 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00065574
CAS-Nummer: 62965-35-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR