N-(tert-Butoxycarbonyl)-L-tert-leucine 98%

Lieferant: Thermo Fisher Scientific

368010010 368010050 368010250
ACRO368010010EA 33.5 EUR
ACRO368010010 ACRO368010050 ACRO368010250
N-(tert-Butoxycarbonyl)-L-tert-leucine 98%

CAS No.: 62965-35-9

Formel: C₁₁H₂₁NO₄
Schmelzpunkt: 118…121 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00065574
CAS-Nummer: 62965-35-9

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Crystalline powder or crystals
Infrared spectrum Conforms
Specific optical rotation -3.0° to -5.0° (20°C, 589 nm) (c=1, acetic acid)
Titration with NaOH ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR