4-Brom-m-kresol 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-Brom-3-methylphenol, 2-Brom-5-hydroxytoluol, 1-Brom-4-hydroxy-2-methylbenzol

368200050
ACRO368200050EA 80.8 EUR
ACRO368200050
4-Brom-m-kresol 98%
4-Brom-m-kresol
Formel: C₇H₇BrO
Molecular Weight: 187,04 g/mol
Siedepunkt: 142…145 °C (23 mmHg)
Schmelzpunkt: 57…61 °C
MDL: MFCD00079723
CAS-Nummer: 14472-14-1
EINECS: 238-464-3

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Crystalline powder and/or chunks
Infrared spectrum Conforms
Melting point 57 to 61 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR