4-Brompiperidinhydrobromid 98%

Lieferant: Thermo Fisher Scientific
Achtung

368310010 368310100
ACRO368310010EA 35.7 EUR
ACRO368310010 ACRO368310100
4-Brompiperidinhydrobromid 98%
4-Bromopiperidine hydrobromide

Appearance: White Crystalline powder

Formel: C₅H₁₁Br₂N
Molecular Weight: 244.9600 g/mol
Schmelzpunkt: 199…202 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00191858
CAS-Nummer: 54288-70-9

Bestellen

Spezifikation Testergebnisse

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Melting point 191 to 196 °C
Titration Argentometric 97.5 to 102.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR