3-Iodbenzonitril 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
368600010
368600100
ACRO368600010EA
13.2
EUR
ACRO368600010
ACRO368600100
3-Iodbenzonitril 97%
3-Iodbenzonitril
Formel:
C₇H₄IN Molecular Weight: 229.02 g/mol Schmelzpunkt: 39…44 °C Flammpunkt: >100 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00079762 CAS-Nummer: 69113-59-3 UN: 3439 ADR: 6.1 III |
Spezifikation Testergebnisse
Appearance | White to brown Powder or crystals and/or chunks |
Infrared spectrum | Conforms |
Melting point | 38 to 44 °C |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...