Tebbe Reagent 0.5 M in Toluol
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
µ-Chloro-µ-methylenebis(cyclopentadienyl)titaniumdimethylaluminum, Bis(cyclopentadienyl-mu-chloro(dimethylaluminium-mu-methylenetitanium
368970250
ACRO368970250EA
254
EUR
ACRO368970250
Tebbe Reagent 0.5 M in Toluol
Tebbe Reagent
CAS No.: 67719-69-1
Formel:
C₁₃H₁₈AlClTi Molecular Weight: 284,62 g/mol Dichte: 0,927 g/cm³ Flammpunkt: 4 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD00151575 CAS-Nummer: 67719-69-1 UN: 2924 ADR: 3,II |
Spezifikation Testergebnisse
Appearance | Dark red to purple Solution |
Strength | 0.40 to 0.60 M |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...