2,3-O-Isopropylidene-D-glyceraldehyde 97%
Lieferant: Thermo Fisher Scientific
Synonyme:
(4R)-2,2-dimethyl-1,3-dioxolane-4-carbaldehyde, (R)-(+)-2,2-Dimethyl-1,3-dioxolane-4-carboxaldehyde, (R)-2,2-dimethyl-1,3-dioxolane-4-carbaldehyde
369260010
ACRO369260010EA
110
EUR
ACRO369260010
2,3-O-Isopropylidene-D-glyceraldehyde 97%
2,3-O-Isopropylidene-D-glyceraldehyde
Formel:
C₆H₁₀O₃ Siedepunkt: 139 °C (760 mmHg) Dichte: 1,045 g/cm³ Flammpunkt: 62 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD00269682 CAS-Nummer: 15186-48-8 |
Spezifikation Testergebnisse
Appearance | Clear to hazy colorless to yellow Liquid or solid |
Infrared spectrum | Conforms |
Additional info | Crack to monomer by distillation before use |
GC | ≥96.0 % |
Specific optical rotation | +39° to +69° (20°C, 589 nm) (C=2, Chloroform) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...