2-Methyl-2-propanyl-N-(diphenylmethylen)glycinat 98%

Lieferant: Thermo Fisher Scientific

Synonyme: N-(Diphenylmethylene)glycine tert-butyl ester, Glycine, N-(diphenylmethylene)-, 1,1-dimethylethyl ester, tert-Butyl 2-((diphenylmethylene)amino)acetate

369320010
ACRO369320010EA 91.7 EUR
ACRO369320010
2-Methyl-2-propanyl-N-(diphenylmethylen)glycinat 98%
tert-Butyl-2-((diphenylmethylene)amino)acetate
Formel: C₁₉H₂₁NO₂
Molecular Weight: 295,382 g/mol
Schmelzpunkt: 111…115 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00134280
CAS-Nummer: 81477-94-3

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 111 to 115 °C
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR