Ethylenebis(triphenylphosphine)platinum(0) 98%

Lieferant: Thermo Fisher Scientific
Achtung

369360010 369362500
ACRO369360010EA 382 EUR
ACRO369360010 ACRO369362500
Ethylenebis(triphenylphosphine)platinum(0) 98%
Ethylenebis(triphenylphosphine)platinum(0)
Formel: C₃₈H₃₄P₂Pt
Lagertemperatur: Kühlschrank
MDL: MFCD00012028
CAS-Nummer: 12120-15-9

Bestellen

Spezifikation Testergebnisse

Appearance Light yellow to beige Powder
Infrared spectrum Conforms
Melting point 127 to 131 °C
Assay ≥97.5 % (ex Pt)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR