Chlor-bis-(cycloocten)-rhodium(I) Dimer 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
369371000
369375000
ACRO369371000EA
58.7
EUR
ACRO369371000
ACRO369375000
Chlor-bis-(cycloocten)-rhodium(I) Dimer 98%
Chlorobis(cyclooctene)rhodium(I) dimer
Formel:
C₃₂H₅₆Cl₂Rh₂ Molecular Weight: 717.51 g/mol Schmelzpunkt: 187…191 °C (Zers.) Lagertemperatur: Kühlschrank |
MDL:
MFCD00013287 CAS-Nummer: 12279-09-3 |
Spezifikation Testergebnisse
Appearance | Orange-brown Powder |
Infrared spectrum | Conforms |
Rhodium (Rh) | 28.0 to 29.4 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...