3,4-Dimethoxyphenylboronsäure 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
369390010
369390050
369390250
ACRO369390010EA
27.8
EUR
ACRO369390010
ACRO369390050
ACRO369390250
3,4-Dimethoxyphenylboronsäure 97%
3,4-Dimethoxyphenylboronsäure
Formel:
C₈H₁₁BO₄ Schmelzpunkt: 238…240 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD01074574 CAS-Nummer: 122775-35-3 |
Spezifikation Testergebnisse
Appearance | White to beige Powder or granules and/or chunks |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...