4-Hydroxyphenylboronsäure 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
369420050
369420010
ACRO369420050EA
187
EUR
ACRO369420050
ACRO369420010
4-Hydroxyphenylboronsäure 97%
4-Hydroxyphenylboronsäure
Formel:
C₆H₇BO₃ Schmelzpunkt: 285 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD01074628 CAS-Nummer: 71597-85-8 |
Spezifikation Testergebnisse
Appearance | Off-white to light brown Powder |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...