Methyl-3-(4-hydroxyphenyl)propanoat 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
3-(4-Hydroxyphenyl)propionic Acid Methyl Ester, Methyl 3-(4-hydroxyphenyl)propanoate, Methyl 3-(4-hydroxyphenyl)propionate, 3-(4-Hydroxy-phenyl)-propionic acid-OMe
369430050
ACRO369430050EA
48.1
EUR
ACRO369430050
Methyl-3-(4-hydroxyphenyl)propanoat 98%
Methyl-3-(4-hydroxyphenyl)propionate
Appearance: Transparent white Low melting mass
Formel:
C₁₀H₁₂O₃ Molecular Weight: 180,20 g/mol Siedepunkt: 108 °C (11 mmHg) Schmelzpunkt: 37…41 °C Flammpunkt: >110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00071577 CAS-Nummer: 5597-50-2 |
Spezifikation Testergebnisse
Appearance | White to beige Crystalline powder or crystals or fused mass |
Infrared spectrum | Conforms |
Melting point | 37 to 41 °C |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...