Natrium-1-decansulfonat 96%

Lieferant: Thermo Fisher Scientific

369450050 369450250 369451000
ACRO369450050EA 64.5 EUR
ACRO369450050 ACRO369450250 ACRO369451000
Natrium-1-decansulfonat 96%
Natrium-1-decansulfonat
Formel: C₁₀H₂₁NaO₃S
Schmelzpunkt: >300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007526
CAS-Nummer: 13419-61-9
EINECS: 236-525-9

Bestellen

Spezifikation Testergebnisse

Appearance White to cream Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤5 % (100°C, 3 h) (vacuum)
Titration after Ion exchange ≥95 % (On dry substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR