Sodium glycocholate hydrate 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: N-Cholylglycine sodium salt, 3α,7α,12α-Trihydroxy-5β-cholan-24-oic acid N-(carboxymethyl)amide sodium salt hydrate, N-[(3α,5β,7α,12α)-3,7,12-Trihydroxy-24-oxocholan-24-yl]-Glycine sodium salt xhydrate, Sodium 2-((4R)-4-((3R,7R,10S,12S,13R,17R)-3,7,12-trihydroxy-10,13-dimethylhexadecahydro-1H-cyclopenta[a]phenanthren-17-yl)pentanamido)acetate hydrate, Glycocholic acid sodium salt hydrate

369460050 369460250
ACRO369460050EA 466 EUR
ACRO369460050 ACRO369460250
Sodium glycocholate hydrate 98%
Sodium glycocholate hydrate

Appearance: White to off-white Powder

Formel: C₂₆H₄₂NO₆Na·xH₂O
Molecular Weight: 487.60 g/mol (anhy)
Schmelzpunkt: 210…215 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09037360
CAS-Nummer: 207614-05-9

Bestellen

Spezifikation Testergebnisse

Appearance White to cream Powder
Infrared spectrum Conforms
Water ≤8 % (K.F.)
HPLC ≥97.5 %
Specific optical rotation On anhydrous basis
Titration with HClO4 97.5 to 102.5 % (On anhydrous basis)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR