Methylthiosalicylat 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Methyl 2-mercaptobenzoate
369600050
369600250
ACRO369600050EA
79.4
EUR
ACRO369600050
ACRO369600250
Methylthiosalicylat 97%
Methylthiosalicylat
Formel:
C₈H₈O₂S Siedepunkt: 98…100 °C (2 mmHg) Flammpunkt: >110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00060678 CAS-Nummer: 4892-02-8 UN: 2810 ADR: 6.1,III |
Spezifikation Testergebnisse
Appearance | Clear colorless to yellow to light green Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5910 to 1.5950 (20°C, 589 nm) |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...