Di-tert-butyliminodicarboxylat 97%

Lieferant: Thermo Fisher Scientific
Achtung

369660050 369660250 369661000
ACRO369660050EA 66.8 EUR
ACRO369660050 ACRO369660250 ACRO369661000
Di-tert-butyliminodicarboxylat 97%
Di-tert-butyliminodicarboxylat
Formel: C₁₀H₁₉NO₄
Schmelzpunkt: 114…117 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00043309
CAS-Nummer: 51779-32-9

Bestellen

Spezifikation Testergebnisse

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR