2-Picolylbromid Hydrobromid 98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 2-(Bromomethyl)pyridine hydrobromide

370110050 370110250 370111000
ACRO370110050EA 59.3 EUR
ACRO370110050 ACRO370110250 ACRO370111000
2-Picolylbromid Hydrobromid 98%
2-Picolylbromid Hydrobromid
Formel: C₆H₇Br₂N
Molecular Weight: 252.94 g/mol
Schmelzpunkt: 149…152 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01863544
CAS-Nummer: 31106-82-8
UN: 2923
ADR: 8 II

Bestellen

Spezifikation Testergebnisse

Appearance White to pink to orange-brown Powder or crystals
Infrared spectrum Conforms
HPLC ≥97.5 %
Titration with HClO4 97.5 to 102.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR