Bis(2,2,2-trifluoroethyl) (methoxycarbonylmethyl)phosphonate 90%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Bis(2,2,2-trifluoroethyl)phosphonoacetic Acid Methyl Ester, Methyl [Bis(2,2,2-trifluoroethoxy)phosphinyl]acetate, Methyl Bis(2,2,2-trifluoroethyl)phosphonoacetate, Methyl O,O'-bis(2,2,2-trifluoroethyl)phosphonoacetate, Methyl 2-(bis(2,2,2-trifluoroethoxy)phosphoryl)acetate, Bis(2,2,2-trifluoroethyl) [(methoxycarbonyl)methyl]phosphonate
370610050
ACRO370610050EA
158
EUR
ACRO370610050
Bis(2,2,2-trifluoroethyl) (methoxycarbonylmethyl)phosphonate 90%
Bis(2,2,2-trifluoroethyl) (methoxycarbonylmethyl)phosphonate
Appearance: Clear orange to dark brown Liquid
Formel:
C₇H₉F₆O₅P Molecular Weight: 318,11 g/mol Dichte: 1,504 g/cm³ Flammpunkt: >110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009901 CAS-Nummer: 88738-78-7 |
Spezifikation Testergebnisse
Appearance | Clear orange to dark brown Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.3700 to 1.3740 (20°C, 589 nm) |
GC | ≥88 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...