1-(4-Methylphenyl)piperazin 98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 1-(p-Tolyl)piperazin

370830050
ACRO370830050EA 80.1 EUR
ACRO370830050
1-(4-Methylphenyl)piperazin 98%
1-(4-Methylphenyl)piperazin
Formel: C₁₁H₁₆N₂
Molecular Weight: 176,26 g/mol
Schmelzpunkt: 29…34 °C
MDL: MFCD00040737
CAS-Nummer: 39593-08-3
EINECS: 254-534-6
UN: 3259

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow Low melting solid
Infrared spectrum Conforms
Melting point 29 to 34 °C
GC ≥97.5 %
Titration with HClO4 ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR