(R)-(+)-3-Boc-2,2-dimethyloxazolidine-4-carboxaldehyde 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: (R)-tert-butyl 4-formyl-2,2-dimethyloxazolidine-3-carboxylate, 3-Oxazolidinecarboxylic acid, 4-forMyl-2,2-diMethyl-, 1,1-diMethylethyl ester, (4R)-, tert-Butyl (R)-(+)-4-formyl-2,2-dimethyl-3-oxazolidinecarboxylate

370890010
ACRO370890010EA 112 EUR
ACRO370890010
(R)-(+)-3-Boc-2,2-dimethyloxazolidine-4-carboxaldehyde 95%
(R)-(+)-3-Boc-2,2-dimethyloxazolidine-4-carboxaldehyde
Formel: C₁₁H₁₉NO₄
Molecular Weight: 229,276 g/mol
Dichte: 1,06 g/cm³
Flammpunkt: 107 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00674064
CAS-Nummer: 95715-87-0

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to light yellow Viscous liquid
Infrared spectrum Conforms
Refractive index 1.4450 to 1.4490 (20°C, 589 nm)
GC ≥94 %
Specific optical rotation +85° to +95° (20°C, 589 nm) (c=1, CHCl3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR