Sand 40-100 mesh, pure
Lieferant: Thermo Fisher Scientific
Synonyme:
Silicium(IV)oxid, Siliciumdioxid
370940010
370940050
370940250
370942500
ACRO370940010EA
81.5
EUR
ACRO370940010
ACRO370940050
ACRO370940250
ACRO370942500
Sand 40-100 mesh, pure
Sand
Formel:
SiO₂ Molecular Weight: 60.08 g/mol Siedepunkt: 2230 °C Schmelzpunkt: 1610 °C Dichte: 2.6 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011232 CAS-Nummer: 14808-60-7 EINECS: 231-545-4 |
Spezifikation Testergebnisse
Appearance | Light beige to grey Crystals |
Additional info | Cr2O3: 0.00005 % |
Particle size | 205 µm (D50) Typical |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...