Hydroxylapatit zur Analyse

Lieferant: Thermo Fisher Scientific

371260250 371261000 371260010
ACRO371260250EA 37.8 EUR
ACRO371260250 ACRO371261000 ACRO371260010
Hydroxylapatit zur Analyse
Hydroxylapatit
Formel: Ca₅HO₁₃P₃
Schmelzpunkt: 1700 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00010904
CAS-Nummer: 1306-06-5
EINECS: 215-145-7

Bestellen

Spezifikation Testergebnisse

Appearance White Powder
Titration Complexometric 35 to 40 % (Ca)
Chloride (Cl) ≤1500 ppm
Fluoride (F) ≤50 ppm
Sulfate (SO4) ≤5000 ppm
Arsenic (As) ≤2 ppm
Copper (Cu) ≤20 ppm
Iron (Fe) ≤400 ppm
Lead (Pb) ≤5 ppm
Mercury (Hg) ≤1 ppm
Zinc (Zn) ≤20 ppm
Heavy metals (as Pb) ≤10 ppm
Insoluble matter ≤0.2 % (in HCl)
Loss on ignition ≤8 % (800°C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR