(Triphenylphosphoranyliden)acetonitril 96%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-(Triphenylphosphoranylidene)acetonitrile, (Cyanomethylene)triphenylphosphorane

371570050
ACRO371570050EA 112 EUR
ACRO371570050
(Triphenylphosphoranyliden)acetonitril 96%
(Triphenylphosphoranylidene)acetonitrile
Formel: C₂₀H₁₆NP
Molecular Weight: 301,33 g/mol
Schmelzpunkt: 189…198 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00567633
CAS-Nummer: 16640-68-9
ADR: 6.1,III

Bestellen

Spezifikation Testergebnisse

Appearance Off-white to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 189 to 198 °C
Additional info The actual assay is mentioned on the COA.
HPLC ≥95 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR