Benzyl-2,5-dihydro-1H-pyrrol-1-carboxylat 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Benzyl 3-pyrroline-1-carboxylate, 1H-pyrrole-1-carboxylic acid, 2,5-dihydro-, phenylmethyl ester, benzyl 2,5-dihydropyrrole-1-carboxylate, Benzyl 2,5-dihydro-1H-pyrrole-1-carboxylate
371690050
ACRO371690050EA
107
EUR
ACRO371690050
Benzyl-2,5-dihydro-1H-pyrrol-1-carboxylat 98%
Benzyl-2,5-dihydro-1H-pyrrole-1-carboxylate
Formel:
C₁₂H₁₃NO₂ Molecular Weight: 203,241 g/mol Siedepunkt: 133…137 °C (,6 mmHg) Dichte: 1,132 g/cm³ Flammpunkt: >110 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD01863559 CAS-Nummer: 31970-04-4 |
Spezifikation Testergebnisse
Appearance | Colorless to light yellow Liquid |
Infrared spectrum | Conforms |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...