2-Fluor-5-nitrobenzaldehyd 97%
Lieferant: Thermo Scientific
|
Achtung
|
371730010
ACRO371730010EA
59.1
EUR
ACRO371730010
2-Fluor-5-nitrobenzaldehyd 97%
2-Fluoro-5-nitrobenzaldehyde
Formel:
C₇H₄FNO₃ Molecular Weight: 169,11 g/mol Schmelzpunkt: 57…60 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00042298 CAS-Nummer: 27996-87-8 |
Spezifikation Testergebnisse
Appearance | Beige to light brown Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 56 to 60 °C |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...