2-Cyanbenzaldehyd 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-Formylbenzonitrile

371860050
ACRO371860050EA 73.2 EUR
ACRO371860050
2-Cyanbenzaldehyd 98%
2-Cyanbenzaldehyd
Formel: C₈H₅NO
Molecular Weight: 131,13 g/mol
Schmelzpunkt: 102…106 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00017503
CAS-Nummer: 7468-67-9
UN: 3439
ADR: 6.1,III

Bestellen

Spezifikation Testergebnisse

Appearance White to beige to orange Powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 102 to 106 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR