2,3-Dihydro-6-isocyano-1,4-benzodioxine 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: N-methylidyne-2,3-dihydrobenzo[b][1,4]dioxin-6-aminium, 2H,3H-benzo[e]1,4-dioxin-6-isocyanide, 6-Isocyano-4-oxachromane

372770010
ACRO372770010EA 124 EUR
ACRO372770010
2,3-Dihydro-6-isocyano-1,4-benzodioxine 95%
2,3-Dihydro-6-isocyano-1,4-benzodioxine
Formel: C₉H₇NO₂
Molecular Weight: 161,16 g/mol
Schmelzpunkt: 81…87 °C
Lagertemperatur: Kühlschrank
MDL: MFCD06200565
CAS-Nummer: 174092-82-1
UN: 2811
ADR: 6.1,III

Bestellen

Spezifikation Testergebnisse

Appearance Yellow to brown Powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 81 to 87 °C
HPLC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR