Mexiletin Hydrochlorid

Lieferant: Thermo Fisher Scientific
Achtung

372910050 372910250 372911000
ACRO372910050EA 29.7 EUR
ACRO372910050 ACRO372910250 ACRO372911000
Mexiletin Hydrochlorid
Mexiletin Hydrochlorid
Formel: C₁₁H₁₇NO
Schmelzpunkt: 202 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00216024
CAS-Nummer: 5370-01-4

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 200 to 205 °C
Loss on drying ≤0.5 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR